missing translation for 'onlineSavingsMsg'
Learn More

Bi-Chromatic Powder

Product Code. 12993531
Encompass
Change view
Click to view available options
Quantity:
2 oz.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12993531 2 oz. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12993531 Supplier Brand Not Defined Supplier No. SSB201L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black/Silver
For Use With (Application) Two-color latent print powder that will contrast with any color surface
Material Bi-Chromatic Powder
Quantity 2 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.