missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Bi-Colour™ 87-900 Series Natural Rubber Latex Immersion Gloves

Product Code. 11722833
Change view
Click to view available options
Size:
7.5
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11722833 11 12 pairs
11731573 7.5 12 pairs
11712833 8 12 pairs
11746883 9 12 pairs
11741573 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11722833 Supplier Ansell™ Supplier No. 87900110BP
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

With a latex/neoprene blend for reliable protection against a very wide range of chemicals. Ansell Bi-Colour™ 87-900 Series Natural Rubber Latex Immersion Gloves are double dipped for double protection.

  • Heavyweight resistance to salts and detergents
  • Quality flocklining in pure cotton to help reduce the risk of skin irritation
  • With embossed lozenge finish, green and yellow in color
  • Strong and durable, with excellent wet or dry grip
  • Ideal for aircraft assembly, battery and electronic manufacture, and the chemical industry
TRUSTED_SUSTAINABILITY

Specifications

Back Material Rubber Latex
Cuff Material Neoprene
Applications Industrial
Coating Material Natural Rubber Latex
Color Blue
Latex Free No
Material Rubber Latex
Product Type Immersion Gloves
Size 11
Thickness (Metric) 0.68 mm
Glove Cuff Color Yellow
Glove Cuff Style Straight
Liner Material Cotton Flocking
Quantity 12 Pairs
Resistant To Salts and detergents resistant
Series 89-900
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.