missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BIBR 1532, TRC
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Specifications
Specifications
| Chemical Name or Material | 2-(((2E)-3-(2-Naphthalenyl)-1-oxo-2-buten-1-yl)amino)-benzoic Acid |
| CAS | 321674-73-1 |
| Molecular Formula | C21H17NO3 |
| Quantity | 50 mg |
| Synonym | BIBR 1532, 2-[[(2E)-3-(2-Naphthalenyl)-1-oxo-2-buten-1-yl]amino]-benzoic Acid |
| InChI Formula | InChI=1S/C21H17NO3/c1-14(16-11-10-15-6-2-3-7-17(15)13-16)12-20(23)22-19-9-5-4-8-18(19)21(24)25/h2-13H,1H3,(H,22,23)(H,24,25)/b14-12+ |
| SMILES | O=C(NC(C=CC=C1)=C1C(O)=O)/C=C(C2=CC(C=CC=C3)=C3C=C2)\C |
| IUPAC Name | 2-[[(E)-3-naphthalen-2-ylbut-2-enoyl]amino]benzoic acid |
| Molecular Weight (g/mol) | 331.36 |
| Formula Weight | 331.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction