missing translation for 'onlineSavingsMsg'
Learn More

Bicifadine Hydrochloride, TRC

Product Code. 30338736
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30338736 10 mg 10mg
30336791 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30338736 Supplier Toronto Research Chemicals Supplier No. TRCB38225010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bicifadine Hydrochloride
CAS 66504-75-4
Molecular Formula C12H15N . HCl
Quantity 10 mg
Synonym 1-(4-Methylphenyl)-3-azabicyclo[3.1.0]hexane Hydrochloride, (+/-)-1-(4-Methylphenyl)-3-azabicyclo[3.1.0]hexane Hydrochloride, CL 220075, MCV 4147, NIH 9542
InChI Formula InChI=1S/C12H15N.ClH/c1-9-2-4-10(5-3-9)12-6-11(12)7-13-8-12;/h2-5,11,13H,6-8H2,1H3;1H
SMILES Cl.Cc1ccc(cc1)C23CNCC2C3
IUPAC Name 1-(4-methylphenyl)-3-azabicyclo[3.1.0]hexane;hydrochloride
Molecular Weight (g/mol) 209.71
Formula Weight 209.0971
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.