missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.3.1]nonane-2,6-dione, 97%, Thermo Scientific™

Product Code. 10623013
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10623013 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10623013 Supplier Thermo Scientific Maybridge Supplier No. BTB12430DA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16473-11-3
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H12O2
MDL Number MFCD00153926
Quantity 1 g
Synonym bicyclo 3.3.1 nonane-2,6-dione, bicyclo 3.3.1 nona-2,6-dione, meerwein's ketone, zlchem 41, acmc-20alon
InChI Key QWNPVTXLBMSEPN-UHFFFAOYSA-N
SMILES C1CC(=O)C2CCC(=O)C1C2
IUPAC Name bicyclo[3.3.1]nonane-2,6-dione
Molecular Weight (g/mol) 152.193
PubChem CID 140084
Formula Weight 152.19
Percent Purity 97%
Chemical Name or Material Bicyclo[3.3.1]nonane-2, 6-dione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.