missing translation for 'onlineSavingsMsg'
Learn More

Bifonazole, 99%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J63253.03

Quantity 1g

 View more versions of this product

Product Code. 15485519

  • 52.00 € / 1g
 Restricted Item
Estimated Shipment: 16-09-2026
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Bifonazole is an imidazole antifungal agent and calmodulin antagonist. It causes a reduction in glycolysis and ATP levels in B16 melanoma cells. Bifonazole is an inhibitor of C14orf1. Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

Solubility
Soluble in DMSO

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

60628-96-8
310.4
OCAPBUJLXMYKEJ-UHFFFAOYSA-N
2378
1-[phenyl-(4-phenylphenyl)methyl]imidazole
C22H18N2
MFCD00865567
Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole
CHEBI:78692
C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4

Specifications

60628-96-8
1g
MFCD00865567
Mycosporan; 1-(alpha,4-Diphenylbenzyl)imidazole
OCAPBUJLXMYKEJ-UHFFFAOYSA-N
1-[phenyl-(4-phenylphenyl)methyl]imidazole
2378
310.39
Bifonazole
White
C22H18N2
14,1213
Soluble in DMSO
C1=CC=C(C=C1)C2=CC=C(C=C2)C(C3=CC=CC=C3)N4C=CN=C4
310.4
CHEBI:78692
99%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.