Learn More
Heinz Herenz Big Foam Swabs Polypropylene Stick
Click to view available options
Unit Size:
Pack of 1000
Description
We offer a wide range of sterile and nonsterile swabs:
- Wood, PS, PP, aluminium or paper applicator
- Each in various lengths available
- With cotton, viscose, polyester or foam head
- Diameter of the heads: small, medium or large
- This guarantees that the right instrument is available for every special application.
- For example, the cotton-tipped swabs are recommended for use by ENT specialists.
- Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
- Only suitable for single use
- Use by medical personnel only
Specifications
Specifications
| Sterility | Non-sterile |
| Applicator Material | Foam |
| Certifications/Compliance | ISO 13485 |
| Handle Material | Polypropylene |
| Shelf Life | 5 Years |
| Tip Material | Foam |
| Product Type | Foam Swab |