Learn More
Trajan Scientific and Medical Big Trap - Oxygen
Product Code. 15790969
Click to view available options
Port Type:
1/4 in.
1/8 in.
Unit Size:
Each
Product Code. 15790969
Brand: Trajan Scientific and Medical 1035314
Only null left
Add to basket
Add to basket
For bulk purification applications or where several instruments are plumbed from a single source, a Big Trap gas filter is an ideal solution. A one-piece heavy walled alminium tube provides 750cm3 of capacity and a pressure rating of up to 250 psig
  • High capacity filters ideal for bulk purification - capacity of 3 L oxygen
  • Efficiency < 1 ppb
  • Can be used where several instruments are plumbed from a single gas source
  • Sintered stainless steel frits prevent particulate contamination
Specifications
For Use With (Application) ECD and TCD when used with a moisture filter, PID when used with a hydrocarbon filter, ICP-AES (ICP-OES) and ICP-MS
Model Oxygen
Volume (Metric) 750 cm3
Port Type 1/4 in.
Description Removes oxygen as well as traces of sulfur and chlorine compounds from carrier gas
For Use With Big Trap Mounting Clip
Pressure 250 psig