missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v1.1 Cycle Sequencing Kit

Product Code. p-7138583 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 Reactions
1000 Reactions
24 Reactions
5000 Reactions
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15869786

Brand: Applied Biosystems™ 4337449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This BigDye™ Terminator v1.1 Cycle Sequencing Kit is designed for specialty applications that require optimal basecalling adjacent to the primer and for sequencing short PCR product templates with rapid electrophoresis run modules.

This BigDye™ Terminator v1.1 Cycle Sequencing Kit is designed for specialty applications that require optimal basecalling adjacent to the primer and for sequencing short PCR product templates with rapid electrophoresis run modules.

  • Improve the quality of your results for a wide range of sequencing applications
  • Sequence challenging templates and sequences more successfully
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs

Tackle Tough Sequences

With the BigDye Terminator v1.1 Cycle Sequencing Kit you get a robust, highly flexible chemistry for a wide range of applications, including de novo sequencing and resequencing.

Get More Uniform Peak Heights, Improved Accuracy

With BigDye Terminator v1.1 kit's superior chemistry, you generate data with uniform peak heights and optimized signal balance-which results in longer, higher-quality reads. You also get more accurate base assignments for heterozygote and mutation detection.

Easily Integrate into Your Workflow

BigDye™ Terminator v.1.1 kit's chemistry requires no new software or instrument recalibration.You can easily integrate either kit into your current workflow with minimal changes to your protocols.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 200 μL Tube of BigDye™ Terminator v1.1 Ready Reaction Mix
• Tube M13 (-21) Primer
• Tube pREF-BDT™ Control DNA (200 ng/μL)
• 1 mL Tube of 5x Sequencing Buffer (may be stored at 4°C)

Store kit at -15°C to -25°C, and avoid repeated freeze-thaw cycles.
For Use With (Equipment) SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐Well and 384‐Well Thermal Cyclers
For Use With (Application) AT-Rich Sequencing (>65%), Comparative Sequencing (Germline Mutations - 50:50), Short PCR Product Sequencing, GC-Rich Sequencing (>65%), GT-Rich or Difficult Template Sequencing, De Novo Sequencing - Low to Mid Throughput
Product Line BigDye™ Terminator
Product Type Sequencing Buffer
Quantity 24 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Plasmid DNA (≤15 Kb), PCR Amplicons, Single Stranded DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.