missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit

Product Code. 15809796 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 Reactions
1000 Reactions
25,000 Reactions
5000 Reactions
Unit Size:
25000 reactions
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15809796 1000 Reactions Each
15781836 100 Reactions Each
15847581 5000 Reactions Each
17152979 25,000 Reactions 25000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15809796 Supplier Applied Biosystems™ Supplier No. 4337456
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Añadir a la cesta

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT-rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs

Get more uniform peak heights and improved accuracy
With BigDye™ Terminator v3.1 kit superior chemistry, you generate data with uniform peak heights and optimized signal balance. This results in longer, higher-quality reads and more accurate base assignments for heterozygote and mutation detection.

TRUSTED_SUSTAINABILITY

Especificaciones

Content And Storage • 10 tubes of BigDye™ Terminator v3.1 Ready Reaction Mix
• M13 (-21) Primer
• pREF-BDT™ Control DNA
• 1 x 12 mL tube of 5X Sequencing Buffer (may be stored at 4°C)

Store kit at -15°C to -25°C and avoid repeated freeze-thaw cycles.
Format 12-well Strips, 8-well Strip, 96- or 384-well Plate
For Use With (Equipment) SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers
For Use With (Application) Ideal for De Novo Sequencing, Resequencing and Finishing with PCR Product, Plasmid, Fosmid and BAC templates
Product Line BigDye™ Terminator
Product Type Sequencing Buffer
Quantity 1000 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Lambda DNA, Genomic DNA (Bacterial), Rolling Circle Amplified Products, Fosmids, PCR Amplicons (50:50 Heterozygotes), Plasmid DNA (≤15 Kb), Single Stranded DNA
What information should I include when submitting data to Genetic Analysis Technical Support?

When contacting Genetic Analysis Support (FAS, FSE, Technical Support), please have (if possible):

Instrument model and serial numbers
Version of the software are you using
Application and/or kit information
Kit lot number
Number of runs on the capillary or array
Polymer type and lot number
Buffer lot number
Hi-Di Formamide lot number or loading solution information
Sample data that can be sent to the support person
Details on:
  -Reaction setup (i.e., how much DNA was used, primer, etc.)
  -How the reactions were cleaned up (alcohol precipitation, columns, etc)
  -What template DNA was used (i.e. plasmid, PCR product)
  -How the template DNA was prepared?

All of this information will help the support person quickly and accurately assess the problem and provide you with recommendations for resolution.

I have a long homopolymer stretch in my sample that I cannot get through. Do you have any tips for getting through it?

In order to get through homopolymeric regions, you can either use anchored sequencing primers or try using the dRhodamine Terminator Cycle Sequencing Kit since it uses ddTTP instead of ddUTP and has been shown to be less prone to producing stutters, specifically with poly-T regions.

For more information on sequencing through homopolymer stretches, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

I am trying to sequence GC-rich DNA without success. Do you have any tips for getting through this?

When trying to sequence through GC-rich regions, the following tips may improve your success:

• Set the reaction up at 1X or 0.5X strength. Heavily diluting the BigDye Terminator Ready Reaction mix can reduce the success of the sequecing reaction through long GC stretches.

• Perform a hot start at 98–99°C for 5 minutes.

• Use 5% (w/v) DMSO or freshly made betaine in the reaction.

• Use the dGTP BigDye Terminator Cycle Sequencing Kit. G peaks will appear compressed due to the presence of ddGTP, but sequencing through long GC stretches using the dGTP kit is typically more successful than with the standard BigDye Terminator Cycle Sequencing Kits (containing ddITP). For more information on sequencing GC-rich DNA, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

I see another sequence running under mine. What can cause this?

Some of the causes of another sequence appearing under the primary sequence are:

• Contamination: There is more than one species of DNA present (e.g., multiple PCR products).

• Primers: Primer is annealing in more than one location on the template, primer dimer, primer degradation or not manufactured properly resulting in N+1 or N-1, carry over from PCR reaction, or primers pairs in a multiplex reaction that are not appropriate for multiplexing (i.e., primers anneal inappropriately).

• Spectral/Matrix: If the raw data signal intensity of the sample is too high or saturated it can exceed the amount of color bleedthrough (or spectral overlap) that the matrix (310) or spectral (3130, 3730, 3500) are removing, resulting in secondary peaks appearing in a very specific pattern (e.g., a red peak always appearing under a green peak). A change in the camera, laser, or optical alignment requires that a new matrix be made or a new spectral calibration be performed.

For more information on more than one sequence or set of peaks in a sequencing run, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

My data seems top heavy. It starts out strong and then gets weaker. What can cause this?

The cause of this is usually an overabundance of template DNA relative to the amount of BigDye Terminator Ready Reaction mix being used in the reaction. This can drive the reaction to incorporate the labeled ddNTPs closer to the 5’ end of the sequence. To balance the signal, either increase the amount of BigDye Terminator Ready Reaction mix being used in the reaction or decrease the concentration of the template DNA.

For more information on other causes of short reads and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

I am seeing dye breakdown/artifacts in my data. What can cause this?

Some of the causes of dye breakdown or artifacts in the data are:

• Oxidation or change in pH: Injecting out of water or old Hi-Di Formamide that has broken down. Also, some cleanup methods can oxidize the dyes.

• Array: An old or contaminated array can cause dye breakdown.

• Arcing: An arcing event on the system can destabilize the blue baseline and—in severe cases—mask it completely.

For more information on other causes of dye breakdown and artifacts in data and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

I am getting shorter reads than I used to. What can cause this?

A change in the read length is usually caused by something that either changes the migration of the DNA or the efficiency of the sequencing reaction. Some of the things that can cause this are:

• Buffer: Buffer may be prepared incorrectly or on the system for too long.

• Polymer: Polymer may have been on the system the system too long, used past the expiration date, frozen/crystallized and thawed, or mixed with water or buffer during capillary fill. There may be a mismatch between polymer type selected in software and what is installed on the system.

• Environment: Instrument may be operating outside environmental specifications in the Site Preparation Guide. You may have poor/improper instrument ventilation, or air flow may be blowing directly on the system.

• DNA quality: RNA/protein contamination may be reducing the sequencing reaction efficiency. If working with PCR product, there may be carryover of primers and/or dNTPs from the PCR reaction, or the sequencing reaction cleanup is incomplete. If using a kit that has beads in it for the purification, beads may be blocking the capillary tip. If using BigDye XTerminator Purification, heating the XTerminator reagent can cause loss of smaller products.

• DNA quantity: There may be too much DNA competing for entry into the capillary with labeled product, or excessive amounts of DNA creating a temporary blockage of the capillary.

For more information on other causes of short reads and how to address each issue, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the catalog number 4305080.

My DNA sequencing data from the capillary electrophoresis system does not look good. Why?

Capillary electrophoresis systems are complex systems, and troubleshooting data quality can be complicated. The biggest contributors to data quality can be divided into these areas: software, chemistry, and instrument performance.

Troubleshooting software
Ensure that the run module you are using and the dye set match the chemistry and instrument setup. For example, if you are running a multicapillary instrument with a 50 cm array on it, the run module should have a “50” in the name. On a multicapillary system, BigDye Terminator v3.1 should run under Dye Set Z, BigDye Terminator v1.1 chemistry should run in Dye Set E (on the ABI PRISM 310 Genetic Analyzer, both chemistries run under Filter Set E, but require different matrices for analysis). The mobility file (DyeSet/Primer) should have the correct instrument, polymer, and chemistry in the name (e.g. KB_3130_POP7_BDTv.3.mob).

Troubleshooting chemistry and instrument performance
•Sequencing Standards: validate instrument
If Sequencing Standards fail, it suggests a possible electrophoresis problem due to array, polymer, buffer, water, septa, plastics, or maybe a hardware failure such as autosampler, laser, camera, etc.

•pREF-BDT Control DNA: validate cycle sequencing and its clean-up
If the pREF-BDT control reactions fail and the Sequencing Standards pass, look into potential problems with the sequencing kit, thermal cycler, and cleanup procedure.
The pREF-BDT Control DNA and M13 (–21) primers needed to run the reactions are included with each BigDye Terminator Cycle Sequencing Kit

•Custom Internal Controls: validate PCR and its clean-up
If the Sequencing Standards and the pGEM controls passed, then look into potential problems with template quantity and quality, primers, PCR reaction and purification.

For more information on troubleshooting your data, please refer to the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: 3rd Edition, which can be downloaded from this link (https://www.thermofisher.com/us/en/home/global/forms/sanger-sequencing-guide-download-form.html).

Can I dilute my BigDye Terminator reactions?

The BigDye Terminator v1.1 and v3.1 chemistries can be diluted, and a 5X Sequencing Buffer is provided with the kit to enable you to do so. However, dilution of the BigDye Terminator may compromise your data integrity depending on the cycle sequencing protocol you are following and the sequence of the particular DNA you are investigating. TheBigDye Direct Cycle Sequencing Kit and the dGTP BigDye Terminator Ready Reaction Kits v1.0 and v3.0 cannot be diluted.

What is a dye blob?

After the cycle sequencing reaction is complete, unincorporated ddNTPs can form complexes that migrate more slowly than individual ddNTPs would. These complexes can interfere with electrokinetic injection, electrophoretic separation, and data analysis. In the electropherogram these can be observed as a region containing multicolored peaks of varying sizes or blobs, depending on the efficiency of the cleanup. In BigDyeTerminator v3.1 reactions, these blobs may appear around 70–80 bp and, in severe cases, 110–120 bp. For BigDye Terminator v1.1, the blobs will either migrate ahead of the data collection time (so they will not be visible) or they may be visible at the very beginning of the run.

What are the different ways to clean up my sequencing reactions?

While there are many kits and methods available to clean up the extension products from a sequencing reaction, the ones that Thermo Fisher Scientific has formally tested and recommends are:

•BigDye XTerminator Purification Kit
•Ethanol precipitation
•Spin column/plate purification (e.g. Centri-Sep columns, DTR Kits from Edge Biosystems)

A list of alternative methods for purification is available in the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C, page 119). The guide can be found by searching the Thermo Fisher Scientific website with the Cat. No. 4305080.

What is the difference between the regular BigDye (Terminator) kits and the BigDye Direct Cycle Sequencing Kit?

BigDye Direct Cycle Sequencing Kit supplies reagents for the complete cycle sequencing workflow, including the PCR reaction, reaction cleanup, and sequencing the PCR product. The post-PCR and cycle sequencing process is combined into a single step, and the workflow requires that the initial PCR primers be designed with an M13 tail, since M13 primers will be used for the sequencing reaction. BigDye Terminator Cycle Sequencing Kits are designed so that the initial PCR and the cleanup are performed as separate reactions. This allows the user more flexibility in primer design and the option to break up the workflow. For the comparative studies that we performed in-house, we found sequencing data obtained using these two kits to be concordant and of at least equivalent quality (typically the BigDye Direct Cycle Sequencing Kit produced better results).

I ran out of 5X Sequencing Buffer. What is the formulation so I can make my own?

The formulation of the 5X Sequencing Buffer that comes with the BigDye Terminator Kit is proprietary.

What is in the BigDye Terminator Ready Reaction Mix?

The BigDye Terminator Ready Reaction Mix contains the dNTPs, fluorescently labeled ddNTPs, buffer, and other components that are needed to drive the sequencing reaction. The specific composition of the BigDye Terminator Ready Reaction Mix is proprietary.

Which kit is best for my application?

Selecting the best kit for your application depends on what you are trying to do—for example, the goals and needs for a resequencing project may be different than for SNP detection. To help guide you, Thermo Fisher Scientific has put together an Applications/Chemistry table found in the DNA Sequencing by Capillary Electrophoresis: Applied Biosystems Chemistry Guide: Second Edition (Cat. No. 4305080, Rev. C). The guide can be found by searching the Thermo Fisher Scientific website with the Cat.No 4305080.

My DNA is X bp. Which BigDye Terminator kit should I use?

The BigDye Terminator v1.1 kit is optimized to produce better incorporation close to the primer and is typically able to produce sequence out to approximately 700–750 bp under optimal conditions (e.g., a well-maintained instrument, clean DNA). The BigDye Terminator v3.1 kit is optimized for longer reads, but the resolution in the first 30–50 bp may be of low quality. Under optimal conditions (e.g., a well-maintained instrument and clean DNA), The BigDye Terminator v3.1 kit is typically able to produce sequence out to approximately 1,200 bp.

What is the difference between the BigDye Terminator v3.1 and v1.1 kits?

The ddNTPs in the two kits are labeled with different dyes that, while spectrally similar, do require a different Dye Set on our multicapillary systems and a different matrix on our single-capillary system. The ratio of dNTP/ddNTPs is also different to allow better incorporation of ddNTPs closer to the primer in the case of the BigDye Terminator v1.1 kit or a longer read in the case of BigDye Terminator v3.1 kit.

What are the concentrations of the pREF-BDT and M13 primer controls that come with the BigDye Terminator kits?

The concentration of the pREF-BDT control is 200 ng/µL. The concentration of the M13 primer is 0.8 pmol/µL.

Should I use the BigDye Terminator v3.1 Cycle Sequencing Kit or BigDye Direct kit if I want to directly perform cycle sequencing on long-range amplicons (up to 10kb) using PCR primers for sequencing, instead of M13 primers?

We recommend the BigDye Terminator v3.1 Cycle Sequencing Kit if you would like to use custom PCR primers instead of M13 primers for sequencing.

The BigDye Direct kit must use the M13 primers provided in the kit for sequencing, as the kit contains the PCR and Sequencing components and are optimized to work together.

Do you offer the BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer from the BigDye Terminator v3.1 Cycle Sequencing Kit as a standalone product?

The BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer is available separately and comes in three volumes, 1mL( SKU 4336697), 28mL ( SKU 4336699), and 233mL ( SKU 4336701).

How do I determine the annealing temperature for the DNA sequencing thermal cycling conditions?

Can I purchase the -21 M13 forward control primer from the BigDye Terminator kit separately?

We do not sell the -21 M13 forward control primer separately, however we provide the primer sequence for your convenience: TGTAAAACGACGGCCAGT

Please feel free to use our oligo services to order this primer (https://www.thermofisher.com/us/en/home/products-and-services/product-types/primers-oligos-nucleotides/invitrogen-custom-dna-oligos.html).
We also offer an -21M13 forward primer that is one basepair shorter than that contained in the kit (Cat. No. N52002).

Can I purchase the pREF-BDT control from the BigDye Terminator kit separately?

We do not sell the pREF-BDT control separately from the DNA sequencing kit.

What is the concentration of the pGEM control and the -21 M13 Control Primer in the BigDye Terminator kit?

The pGEM control is 200 ng/µL and the M13 primer 0.8 pmol/µL

What is the control in the BigDye Terminator kit?

The control is pREF-BDT control plasmid DNA which serves as a double-stranded control

How do I calculate the amount of BigDye Terminator v1.1 & v3.1 5X Buffer to use in the reaction?

The calculation can be done manually or by using our Applied Biosystems BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer Calculator.

The formula is provided below:

Volume of BigDye Terminator v1.1 & v3.1 5X Buffer (µL) = 0.5 x [(total reaction volume/2.5) - volume of BigDye Terminator Ready Reaction Mix].

For example, if you are using 2 µL of the BigDye Terminator Ready Reaction Mix and a 15 µL reaction volume

Volume of BigDye Terminator v1.1 & v3.1 5X Buffer (µL) = 0.5 x [(15/2.5) - 2] = 2 µL

Can I dilute my BigDye Terminator reaction?

We provide the BigDye Terminator v1.1 & v3.1 5X Buffer to enable dilution of the BigDye Terminator Ready Reaction Mix. However, without optimization, diluting the BigDye Terminator Ready Reaction Mix may cause deterioration of sequencing quality. We cannot guarantee the performance of BigDye chemistry when it is diluted.

What is the purpose of the BigDye Terminator v1.1 & v3.1 5X Buffer in the BigDye Terminator kits?

The BigDye Terminator v1.1 & v3.1 5X Buffer is used to maintain the optimal buffer concentration in the sequencing reaction when less than the recommended volume of BigDye Terminator Ready Reaction Mix is used. The BigDye Terminator v1.1 & v3.1 5X Buffer amount varies depending on the volume of the BigDyeTerminator Ready Reaction Mix. To determine the amount of BigDye Terminator v1.1 & v3.1 5X Buffer, please use the Applied Biosystems BigDye Terminator v1.1 & v3.1 5X Sequencing Buffer Calculator.

How much primer should I use in a DNA sequencing reaction?

The primer amount in the final reaction should be 3.2 pmoles. If the sequencing reaction is being diluted, the primer concentration may need additional optimization.

What level of purification do I need for my DNA sequencing primers?

We recommend using HPLC-purified primers for DNA sequencing. This will ensure the presence of full-length, highly purified primers that will minimize cycle sequencing noise and provides longer sequencing reads.

When should I use the BigDye Terminator v3.1 Cycle Sequencing Kit?

If longer read lengths are needed, the BigDye Terminator v3.1 Cycle Sequencing kit has been formulated to deliver robust performance across a wide variety of DNA sequences while maximizing read lengths.

What are the differences between the BigDye Terminator v1.1 Cycle Sequencing Kit and the BigDye Terminator v3.1 Cycle Sequencing Kit?

The BigDye Terminator kit versions have different fluorescent dyes attached to the dideoxy terminators, and the dNTP/ddNTP ratio differs between the kits. The BigDye Terminator v1.1 Cycle Sequencing Kit is optimized for shorter fragments and for obtaining sequence close to the primer. The BigDye Terminator v3.1 Cycle Sequencing Kit is more suitable for larger templates and for obtaining longer read lengths.

What is offscale sequencing data?

Offscale data refers to data from samples that have signal intensities resulting in saturation of the CCD camera, on the capillary electrophoresis instruments. The maximum signal thresholds for raw data are 32,000 rfu for the Applied Biosystems 3730/3730xl DNA Analyzers, 3500/3500xL Genetic Analyzers, and SeqStudio Genetic Analyzer, and 8,000 rfu for the Applied Biosystems 310 Genetic Analyzer and Applied Biosystems 3130/3130xl Genetic Analyzers. For the SeqStudio Flex Genetic Analyzers, if the offscale recovery (OSR) is on, the maximum signal threshold is 65,000 rfu. If OSR is turned off, the maximum signal threshold for the SeqStudio Flex Genetic Analyzers is 32,000 rfu.

What are M13-tailed primers used for in DNA sequencing and can I have the sequence for these primers?

The M13-tailed primers are used to simplify the workflow when sequencing PCR products and they reduce the loss of the 5' unresolvable bases. When the PCR primers contain M13 tails on their 5' ends, the M13 sequence is incorporated into the amplicons. This enables the use of sequencing master mixes containing either the universal M13 forward or M13 reverse primers. The sequence for the M13 forward and reverse primers are as follows:

-M13 forward primer sequence: 5′ TGTAAAACGACGGCCAGT 3′
-M13 reverse primer sequence: 5′ CAGGAAACAGCTATGACC 3′

What is the Primer Designer Tool?

Primer Designer Tool is a free online tool to search for the appropriate PCR/Sanger primer pair from a database of >650,000 pre-designed primer pairs for resequencing the human exome. For more information, including a direct link to purchase the designed primers online go here (http://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/pre-designed-primers-pcr-sanger-sequencing.html?CID=primerdesigner).

Are there any tools that can assist with primer design for DNA sequencing?

We have the Primer Designer Tool, which is a free online tool to search for the appropriate PCR/Sanger primer pair from a database of >650,000 pre-designed primer pairs for resequencing the human exome. Go here (http://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/pre-designed-primers-pcr-sanger-sequencing.html?CID=primerdesigner) for more information, including a direct link to purchase the designed primers online.

How often should I run the pGEM control?

The pGEM control and M13 primer provided in the kit should be used for troubleshooting purposes. The pGEM is a control template that can be used to isolate issues with sample quality, thermal cycler, kit or sequencing reaction purification.

How much primer should I use in my DNA sequencing reaction?

We recommend a primer amount of 3.2 pmoles in the sequencing reaction.

What is the best way to store my purified DNA sequencing reaction?

For long-term storage, the purified sequencing reaction should be dried and stored at -20 degrees C, protected from light.

What are appropriate stopping points after cycle sequencing and for how long/what temperature?

- After the sequencing reaction, post thermal cycling, the samples can be stored at 4 degrees C overnight or at -15 degrees C or -25 degrees C for long-term storage.
- After purifying with Centri-Sep or ethanol precipitation, dry down the samples, seal the plate with MicroAmp Clear Adhesive Film, and store, protected from light, at 4 degrees C for capillary electrophoresis (CE) preparation or at -20 degrees C until use.
- After BigDye Xterminator purification, to store for up to 10 days, seal the plate with MicroAmp Clear Adhesive Film, and store at 4 degrees C for CE preparation or at -20 degrees C. BDX plates can be stored at room temperature for up to 48 hours inclusive of time on the CE instrument.

For other DNA sequencing purification methods, please refer to the reagent-specific protocol.

What template purity is needed for DNA sequencing?

Template quality impacts the DNA sequencing reaction. We recommended a spectrophotometer to determine DNA quality. Optimum absorbance ratios (A260/280) are between 1.8 and 2.0.

Can I directly sequence RNA?

RNA cannot be sequenced directly; instead we recommend transcribing the RNA to cDNA. The cDNA can then be used as a template for sequencing. For more information on recommended kits to convert RNA to cDNA go here (https://www.thermofisher.com/us/en/home/life-science/pcr/reverse-transcription.html.html?icid=fr-ssiii-main).

Since DNA sequencing is similar to PCR, can I multiplex my sequencing reactions?

The Sanger sequencing reaction cannot be multiplexed, as the reaction should contain only one template and one primer. Multiple priming sites will result in sequencing noise and low quality data.

Where can I get additional information on DNA sequencing?

Our website provides many resources for obtaining information regarding sequencing applications. A few are listed below:

-Sanger Sequencing Overview (https://www.thermofisher.com/us/en/home/life-science/sequencing/sanger-sequencing/applications.html)
-DNA Sequencing by Capillary Electrophoresis Guide (http://tools.thermofisher.com/content/sfs/manuals/cms_041003.pdf)
-Seq It Out Video Series (https://www.thermofisher.com/us/en/home/life-science/sequencing/sequencing-education/seq-it-out.html)
-For additional resources, please see the Guides and Tools section on this page (http://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/capillary-electrophoresis-applications-support-center/sanger-sequencing-support.html).

What is the difference between Sanger sequencing and fragment analysis?

DNA sequencing is the determination of the base-pair sequence of a DNA fragment by the formation of extension products of various lengths. DNA sequencing on capillary electrophoresis platforms is the process of reading nucleotide bases in a DNA molecule. During Sanger sequencing, DNA polymerases copy single-stranded DNA templates by adding nucleotides to a growing chain (extension product). This method involves copying single-stranded DNA with chemically altered bases called dideoxynucleotides which, when incorporated at the 3' end of the growing chain, terminate the chain selectively at A, C, G, or T. The terminated chains are then resolved by capillary electrophoresis.

Fragment analysis determines the relative size of a DNA fragment, typically a PCR product. One of the PCR primers is fluorescently labeled and the labeled PCR product is combined with a size standard to extrapolate the base-pair size of the sample.


For Research Use Only. Not for use in diagnostic procedures.

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.