missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye™ Terminator v3.1 Cycle Sequencing Kit

Product Code. 15847581 Shop All Applied Biosystems Products
Click to view available options
Quantity:
100 Reactions
1000 Reactions
25,000 Reactions
5000 Reactions
Unit Size:
25000 reactions
Each
This item is not returnable. View return policy

Product Code. 15847581

Brand: Applied Biosystems™ 4337457

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

The BigDye™ Terminator v3.1 Cycle Sequencing Kit's robust, highly flexible chemistry is ideal for de novo sequencing, resequencing, and finishing with PCR product, plasmid, fosmid, and BAC templates.

  • Improve the quality of your results for a wide range of sequencing applications
  • Optimized for long read lengths
  • Better dye mobility characteristics
  • Improved performance reading through GT-rich regions
  • Get longer, higher-quality reads with more uniform peak heights and optimal signal balance
  • Enhance your productivity and reduce costs

Get more uniform peak heights and improved accuracy

With BigDye™ Terminator v3.1 kit superior chemistry, you generate data with uniform peak heights and optimized signal balance. This results in longer, higher-quality reads and more accurate base assignments for heterozygote and mutation detection.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 x 20 mL tubes of BigDye™ Terminator v3.1 Ready Reaction Mix
• 2 tubes M13 (-21) Primer
• 2 tubes pREF-BDT™ Control DNA
• 2 x 28 mL tubes of 5X Sequencing Buffer (may be stored at 4°C)

Store kit at -15°C to -25°C and avoid repeated freeze-thaw cycles.
Format 12-well Strips, 8-well Strip, 96- or 384-well Plate
For Use With (Equipment) SeqStudio™ Genetic Analyzer, SeqStudio™ Flex Series Genetic Analyzer, 3500/3500xL Genetic Analyzer, 3730/3730xl DNA Analyzer Thermal Cyclers: GeneAmp™ PCR System 9700 Dual Series, Veriti™ Fast 96‐well and 384‐well Thermal Cyclers
For Use With (Application) Ideal for De Novo Sequencing, Resequencing and Finishing with PCR Product, Plasmid, Fosmid and BAC templates
Product Line BigDye™ Terminator
Product Type Sequencing Buffer
Quantity 5000 Reactions
Technique Fluorescent Dye Terminator Sequencing
Template Compatibility BAC DNA, Plasmid DNA (≤15Kb), PCR Amplicons, Single Stranded DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.