missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ BigDye XTerminator™ Purification Kit

Product Code. 15879816 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
2500 Preps
40,000 Preps
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15879816 2500 Preps Each
15889816 40,000 Preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15879816 Supplier Applied Biosystems™ Supplier No. 4376484

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Includes

50 mL XTerminator™ solution (store at 4°C), 225 mL SAM™ solution (store at room temperature or 4°C)

The BigDye XTerminator™ Purification Kit is a fast, simple purification method for DNA sequencing reactions that removes unincorporated BigDye™ terminators and salts.

  • Eliminate liquid transfer - Use a single plate
  • Obtain complete dye blob removal
  • Stabilize the samples before analysis
  • Improve your sequencing workflow
  • Perform manual or automated protocols
  • Get rapid, reliable, and reproducible results

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 50 mL XTerminator™ Solution (store at 4°C), 225 mL SAM™ Solution (store at room temperature or 4°C)
Format Kit
Sample Type Sequencing Products
Final Product Type DNA
For Use With (Application) Sequencing
Includes 2 Bottles
No. of Reactions 2500
Product Line BigDye XTerminator, BigDye
Product Type Purification Kit
Quantity 2500 Preps
Target Sequencing Reactions
Test Time 40 min
Starting Material Amount 10 μl
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • BTL XTERMINATOR SOLUTION 800ML
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
Faresætning
  • H317-May cause an allergic skin reaction.
Sikkerhedssætning
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplerende oplysninger
  • MIXTURE LIST-Contain: DOWEX MR-3 MIXED IONEXCHANGE RESIN

For Research Use Only. Not for use in diagnostic procedures.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.