missing translation for 'onlineSavingsMsg'
Learn More

Biguanide Hydrochloride, TRC

Product Code. 30240826
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240826 5 g 5g
30240727 1 g 1g
30240610 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240826 Supplier Toronto Research Chemicals Supplier No. TRCB0165565G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4761-93-7
Molecular Formula C2H7N5 . HCl
Molecular Weight (g/mol) 137.57
Synonym Diguanide Hydrochloride, Imidodicarbonimidic Diamide Hydrochloride, Guanylguanidine Hydrochloride, Isobiguanide Hydrochloride, (Aminoiminomethyl)-guanidine Hydrochloride;
IUPAC Name 1-(diaminomethylidene)guanidine;hydrochloride
SMILES Cl.NC(=NC(=N)N)N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H7N5.ClH/c3-1(4)7-2(5)6;/h(H7,3,4,5,6,7);1H
Formula Weight 137.0468
Chemical Name or Material Biguanide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.