missing translation for 'onlineSavingsMsg'
Learn More

Bilirubin, Thermo Scientific™

Product Code. 15805098 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Plastic bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15805098 1 g Plastic bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15805098 Supplier Thermo Scientific Supplier No. J12110.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 635-65-4
Packaging Plastic bottle
Molecular Formula C33H36N4O6
MDL Number MFCD00005499
Quantity 1 g
Beilstein 74376
Sensitivity Light sensitive
Synonym bilirubin, hematoidin, hemetoidin, bilirubin ix-alpha, principal bile pigment, unii-rfm9x3lj49, bilirubin ixalpha, 21h-biline-8,12-dipropanoic acid, 2,17-diethenyl-1,10,19,22,23,24-hexahydro-3,7,13,18-tetramethyl-1,19-dioxo, rfm9x3lj49, biline-8,12-dipropionic acid, 1,10,19,22,23,24-hexahydro-2,7,13,17-tetramethyl-1,19-dioxo-3,18-divinyl
InChI Key BPYKTIZUTYGOLE-IFADSCNNSA-N
SMILES CC1=C(C=C)\C(NC1=O)=C\C1=C(C)C(CCC(O)=O)=C(CC2=C(CCC(O)=O)C(C)=C(N2)\C=C2/NC(=O)C(C=C)=C2C)N1
IUPAC Name 3-[2-[[3-(2-carboxyethyl)-5-[(Z)-(3-ethenyl-4-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-2-yl]methyl]-5-[(Z)-(4-ethenyl-3-methyl-5-oxopyrrol-2-ylidene)methyl]-4-methyl-1H-pyrrol-3-yl]propanoic acid
Molecular Weight (g/mol) 584.67
PubChem CID 5280352
ChEBI CHEBI:16990
Formula Weight 584.66
Physical Form Fine Powder
Chemical Name or Material Bilirubin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.