Learn More
Emma Safety Footwear Billy Safety Shoes
Product Code. 15978951
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
49
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15978951 45 1 pair
15918951 39 1 pair
15928951 40 1 pair
15938951 41 1 pair
15948951 42 1 pair
15958951 43 1 pair
15968951 44 1 pair
15988951 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15978951 Supplier Emma Safety Footwear Supplier No. MM73084845
  • ONLINE EXCLUSIVE PRICE
    147.00 €
    92.30 € / 1 pairs
    Save 54.70 €
    37% Off
View all promotions
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
All purpose safety boot repels acid and oil like an umbrella repels water. Strong, ankle supporting upper fits the foot snugly and comfortably.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design Billy
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 45
Toe Material Steel