missing translation for 'onlineSavingsMsg'
Learn More

Bimatoprost Acid, TRC

Product Code. 17982965
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17982965 5 mg 5mg
17934645 25 mg 25mg
17973065 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982965 Supplier Toronto Research Chemicals Supplier No. TRCB3868205MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38344-08-0
Molecular Formula C23 H32 O5
Molecular Weight (g/mol) 388.5
Synonym 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]cyclopentyl]-, (5Z)-, 5-Heptenoic acid, 7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-pentenyl]cyclopentyl]-, (5Z)- (9CI), 5-Heptenoic acid, 7-[3,5-dihydroxy-2-(3-hydroxy-5-phenyl-1-pentenyl)cyclopentyl]-, [1R-[1α(Z),2β(1E,3S*),3α,5α]]-, (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(1E,3S)-3-hydroxy-5-phenyl-1-penten-1-yl]cyclopentyl]-5-heptenoic acid, (5Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(S,E)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]hept-5-enoic acid, 17-Phenyl-18,19,20-trinor-PGF2α, 17-Phenyl-18,19,20-trinorprostaglandin F2α, Bimatoprost acid, PhXA 70, U 35687
IUPAC Name (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(E,3S)-3-hydroxy-5-phenylpent-1-enyl]cyclopentyl]hept-5-enoic acid
SMILES O[C@@H](CCc1ccccc1)\C=C\[C@H]2[C@H](O)C[C@H](O)[C@@H]2C\C=C/CCCC(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H32O5/c24-18(13-12-17-8-4-3-5-9-17)14-15-20-19(21(25)16-22(20)26)10-6-1-2-7-11-23(27)28/h1,3-6,8-9,14-15,18-22,24-26H,2,7,10-13,16H2,(H,27,28)/b6-1-,15-14+/t18-,19+,20+,21-,22+/m0/s1
Formula Weight 388.225
Percent Purity >95%
Chemical Name or Material Bimatoprost acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.