missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BINA
GREENER_CHOICE

Product Code. 18076052
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18076052 50 mg 50mg
18015122 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18076052 Supplier Tocris Bioscience™ Supplier No. 4048/50
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective positive allosteric modulator of mGlu2

Selective positive allosteric modulator of mGlu2 (EC50 = 33.2 nM in CHO cells expressing human mGlu2). Displays no effect on glutamate-induced activation of other mGlu receptor subtypes. Exhibits antipsychotic and anxiolytic properties in mice.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 866823-73-6
Molecular Formula C30H30O4

Specifications

Quantity 50 mg
Formula Weight 454.56
Purity >98%
Molecular Weight (g/mol) 454.566
Chemical Name or Material BINA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.