missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Binding Buffer BB

Product Code. 15839087
Change view
Click to view available options
Quantity:
110 mL
Unit Size:
110mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839087 110 mL 110mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839087 Supplier Macherey-Nagel™ Supplier No. 740394.110
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Binding Buffer BB to be used with NucleoSpin cfDNA XS kits

For rapid purification of circulating DNA from plasma and serum

  • High recovery even of fragmented DNA > 50 bp
  • Elution in as little as 5 μL
  • Concentrated DNA even from dilute samples
  • DNA ready to use for real-time PCR
  • Ideal for analysis of fetal DNA from maternal plasma
  • Ideal for detection of tumor markers in plasma
  • Utilizes mini spin columns with silica membrane technology

Specifications

Chemical Name or Material Buffer BB
CAS 593-84-0
Color Colorless
For Use With (Application) Purification of circulating DNA from plasma or serum
For Use With (Equipment) NucleoSpin™ Plasma XS Kit
pH 5.0 to 7.0
Physical Form Liquid
Quantity 110 mL
Formulation Guanidinium thiocyanate, Ethanol
Odor Odorless
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.