missing translation for 'onlineSavingsMsg'
Learn More

BioAcryl-P (40%, 19:1)

Product Code. 15423599
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423599 500 mL 500mL
15433599 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423599 Supplier Thermo Scientific Alfa Aesar Supplier No. J60909.AP
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

BioAcryl-P (Acrylamide/Bisacrylamide 29:1, CAS # 25034-58-6) is a powder-based formula used to create the gel matrix necessary for electrophoresis experiments.

Acrylamide/Bisacrylamide premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • BioAcryl-P is a 29:1 ratio of Acrylamide/Bisacrylamide
  • Acrylamide is a crystalline amide that forms polymer structures at room temperature
  • Acrylamide acts as the monomer for the gel structure, and bisacrylamide as the crosslinker for gel matrix formation

Application

  • Suitable for gel electrophoresis-mediated separation of proteins and DNA for identification or sequencing
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acrylamide/Bisacrylamide 19:1
Physical Form Liquid
Concentration 40% soln
Color Colorless
Quantity 500 mL
MDL Number MFCD00080848
UN Number UN3426
Solubility Information It is soluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.