missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BioAcryl-P (40%, 39:1)
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Description
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.
Solubility
Fully miscible in water.
Notes
Protect from heat. Store away from oxidizing agents.
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.
Solubility
Fully miscible in water.
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | Acrylamide/Bisacrylamide 39:1 |
| Physical Form | Liquid |
| Concentration | 40% soln |
| Color | Colorless |
| Quantity | 1 L |
| UN Number | UN3426 |
| Solubility Information | Fully miscible in water. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction