Learn More
Thermo Scientific™ BioBasic™ PREP SEC 300 Size Exclusion Chromatography HPLC Columns
Outstanding size separation of water soluble compounds.
Brand: Thermo Scientific™ 73505-307846
Code : 50
Additional Details : Weight : 0.20000kg
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 300 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 7,000 to 1,200,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 300Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
High Temperature Column | |
300 mm | |
5 μm | |
L59 | |
BioBasic | |
Size Exclusion | |
High Temperature Column |
7.8 mm | |
5800 psi (400 bar) | |
300 Å | |
Diol (OH) | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.