Learn More
Thermo Scientific™ BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns
Achieve outstanding size separation of water soluble compounds with BioBasic™ PREP SEC 1000 Size Exclusion Chromatography HPLC Columns.
681.53 € - 2686.85 €
Specifications
Diameter (Metric) | 7.8 mm |
---|---|
For Use With | Preparative Size Exclusion |
Max. Pressure | 4300 psi (295 bar) |
Particle Size | 5 μm |
pH | 2 to 8 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
16691674
|
Thermo Scientific™
73605-037821A |
5 μm | 7.8 mm | 30 mm |
681.53 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||
16601684
|
Thermo Scientific™
73605-157846A |
5 μm | 7.8 mm | 150 mm |
1571.90 €
Each |
In Stock
Log in to see stock. |
|||
16546130
|
Thermo Scientific™
73605-307846A |
5 μm | 7.8 mm | 300 mm |
2686.85 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 1000 Size Exclusion Chromatography preparative HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 70,000 to 10,000,000 daltons, allowing for the analysis of proteins, peptides and other biomolecules.
BioBasic™ PREP SEC 1000 HPLC Column Benefits:
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 1000Å pore size
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
7.8 mm | |
4300 psi (295 bar) | |
2 to 8 | |
60°C | |
Diol (OH) | |
>0.6% | |
No |
Preparative Size Exclusion | |
5 μm | |
25 m²/g | |
L59 | |
Spherical, Fully Porous, Ultrapure Silica | |
Size Exclusion | |
Size Exclusion |