Learn More
Nitritex™ BioClean-D Disposable Laboratory Coats
Product Code. 12942255
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 30
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12942255 X-Large Pack of 30
12912255 Small Pack of 30
12922255 Medium Pack of 30
12932255 Large Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12942255 Supplier Nitritex™ Supplier No. BDLCXL
  • ONLINE EXCLUSIVE PRICE
    295.10 €
    173.57 € / Pack of 30
    Save 121.53 €
    41% Off
View all promotions
In Stock
Add to basket
Add to basket
Wear Nitritex™ BioClean-D Disposable Laboratory Coats to protect against laboratory contamination.
  • Open cuff style
  • Three pockets
  • CleanTough™ Fabric
  • Disposable, packaged in bag
  • Spunbond Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size X-Large
Material Double Laminate Microporous Fabric
Color White
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Size (Metric) Chest 1066.8 to 1143 mm
Certifications/Compliance ISO 9001:2015, ASTM F739-12
Certifications/Compliance (Europe) (EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2
Packaging Type One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 30 outer bags per lined carton (30 pieces)
Construction Spunbond Polypropylene