missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean - D Sterile Kit S BDKM - Size 3XL
BIOPHARMA

Product Code. 17680767
Change view
Click to view available options
Product Type:
Cleanroom Coveralls
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17680767 Cleanroom Coveralls Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17680767 Supplier Ansell Supplier No. SBDKM3XL
  • ONLINE EXCLUSIVE PRICE
    307.00 €
    254.77 € / Pack of 12
    Save 52.23 €
    17% Off
Estimated Shipment: 24-06-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile coverall featuring a collar, hood, overboots and integrated face mask. Hood with integrated face mask with added product and personal protection. Single kit for all garments resulting in less overall packaging waste. Slip-resistant sole for reduced risk of workplace injuries.

  • Added features: The BioClean-D Sterile Kit S-BDKM features a disposable coverall, overboots with a slip-resistant sole, a hood and an integrated face mask, delivering head-to-toe personal protection
  • Optimized packaging: All garments are presented in a single package, making handling more convenient and minimizing overall packaging waste
  • Enhanced fit: The BioClean™ coverall included in this kit is designed with a front zip, elasticated back, cuffs, and ankles, offering a comfortable fit, while thumb loops ensure a secure hold
  • Reduced contamination risks: As it is made from CleanTough™ material, which is both lightweight and low-linting, this sterile coverall is ideal for cleanroom use
TRUSTED_SUSTAINABILITY

Spécification

Product Type Cleanroom Coveralls
Size 3X-Large
Gender Unisex
Material CleanTough
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Chemicals, Particle
Back Style Elasticated
Certifications/Compliance (Europe) CE 0598, EN 1149-5:2008, EN 13935-2, EN 13034:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, ISO 11137-1:2006, ISO 9001:2015, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009
No. of Pockets None
Height (Metric) 194 -200 cm (6 ft. 4 in. - 6 ft. 6 in.)
Cleanroom Class Class 10 (ISO 4)
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.