missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean - D Sterile Kit S BDKM - Size M
BIOPHARMA

Product Code. 17666745
Change view
Click to view available options
Product Type:
Cleanroom Coveralls
Unit Size:
Pack of 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
17666745 Cleanroom Coveralls Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17666745 Supplier Ansell Supplier No. SBDKMM
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Estimated Shipment: 04-05-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile coverall featuring a collar, hood, overboots and integrated face mask. Hood with integrated face mask with added product and personal protection. Single kit for all garments resulting in less overall packaging waste. Slip-resistant sole for reduced risk of workplace injuries.

  • Added features: The BioClean-D Sterile Kit S-BDKM features a disposable coverall, overboots with a slip-resistant sole, a hood and an integrated face mask, delivering head-to-toe personal protection
  • Optimized packaging: All garments are presented in a single package, making handling more convenient and minimizing overall packaging waste
  • Enhanced fit: The BioClean™ coverall included in this kit is designed with a front zip, elasticated back, cuffs, and ankles, offering a comfortable fit, while thumb loops ensure a secure hold
  • Reduced contamination risks: As it is made from CleanTough™ material, which is both lightweight and low-linting, this sterile coverall is ideal for cleanroom use
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Gender Unisex
Material Polyethylene, Non-woven Polypropylene Laminate
Color White
Disposable Single-use
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Standard, Elastic Face
Closure Type Zipper
Seam Type Bound
Sterility Sterile
Resistance Chemicals, Particle
Back Style Elasticated
Certifications/Compliance (Europe) CE 0598, EN 1149-5:2008, EN 13935-2, EN 13034:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, ISO 11137-1:2006, ISO 9001:2015, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009
No. of Pockets None
Height (Metric) 170 - 176 cm (5ft. 6 in. - 5 ft. 9 in.)
Cleanroom Class Class 10 (ISO 4)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.