missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Indigo™ Sterile Nitrile Gloves

Product Code. 15218218
Change view
Click to view available options
Size:
10
5, 5.5
6, 6.5
7, 7.5
8, 8.5
9
Unit Size:
200 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15218218 10 200 pairs
15208218 9 200 pairs
15268208 5, 5.5 200 pairs
15278208 6, 6.5 200 pairs
15288208 7, 7.5 200 pairs
15298208 8, 8.5 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15218218 Supplier Nitritex™ Supplier No. BNPLS1010
Add to basket
Add to basket
This item is not returnable. View return policy

Feature textured fingers for enhanced grip. Nitritex™ BioClean Indigo™ Sterile Nitrile Gloves are latex free gloves that are processed to ensure ISO Class 4 compatibility, are resistant to a wide range of chemicals and offer excellent antistatic properties.

  • Resistant to a wide range of chemicals
  • Textured fingers for enhanced grip
  • Powder-free and latex-free
  • Excellent antistatic properties
  • Beaded cuff
  • Non-particulating EasyTear™ packaging
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) ISO 9001:2015 and (EU)2016/425, EN420:2003 + A1:2009, EN374-1:20016, EN374-5:2016 and EN421:2010, EN455-2:2015, ISO 11137-1:2006, (EC)1935/2004 and (EC)2023/2006, ASTM D6978-05
Cleanroom Class Class 10 (ISO 4)
Color Purple
Disposable Single-use
Exterior Finish Fingertip Textured
Grade EU GMP Grade A
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 10
Sterility Sterile
Glove Cuff Style Beaded
Model BNPLS1010
Packaging Type EasyTear™
Quantity One pair per inner PE wallet; one wallet per sealed EasyTear PE pouch; 10 pouches per sealed outer PE bag; 20 outer bags per lined carton (200 pairs)
Resistant To Chemicals
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.