Learn More
Ansell BIOCLEAN™ Permaflow Irradiated Cleanroom Ballpoint Pen S-BPBP-1
Description
- BioClean Permaflow cleanroom pens have fade and water resistant fast drying permanent ink, and feature an alcohol resistant label for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
- With a robust, lightweight, tinted polypropylene round barrel and 1 mm point size for quality writing precision. Ink complies with documentary use standard ISO 12757-2
Specifications
Specifications
| Tip Style | Rollerball |
| Product Type | Ballpoint Pen |
| Color | Black |