missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens

Product Code. 15278308
Click to view available options
Color:
Black
Blue
Red
Description:
Bagged
Double bagged
Double bagged, Pack of 10
Unit Size:
Pack of 10
Pack of 200
This item is not returnable. View return policy

Product Code. 15278308

Brand: Nitritex™ SBPFPBL1

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.

  • Super permanent ink in black, blue, or red
  • Fade and water resistant
  • Quick drying
  • Non-toxic
  • Alcohol resistant labeling
  • Autoseal ink prevents drying
  • Store in a dry, cool place away from direct sunlight and fluorescent lighting
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Processed in an NEBB certified ISO Class 4 cleanroom environment
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Description Double bagged
Length (English) 5.5 in.
Length (Metric) 140 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.