Learn More
Ansell BIOCLEAN™ Permaflow Sterile Cleanroon Pen S-BPFP-RD1
Product Code. 30040336
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 200
Product Code. Color Unit Size
30040336 Red Pack of 200
1 options
Product Code. 30040336 Supplier Ansell Supplier No. SBPFPRD1
  • ONLINE EXCLUSIVE PRICE
    803.30 €
    484.72 € / Pack of 200
    Save 318.58 €
    39% Off
Estimated Shipment: 23-06-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Ballpoint pens for alcohol resistant labelling are processed to ensure ISO Class 4 and EU GMP Grade A compatibility, fade and water resistant, quick drying, and non-toxic.
  • BioClean Permaflow Cleanroom Pens are sterile with fade and water resistant super permanent ink.
  • With alcohol resistant labeling for easy cleaning within a critical environment, they are ISO Class 4 and EU GMP Grade A compatible.
TRUSTED_SUSTAINABILITY
Specifications
Tip Style Fine
Product Type Permanent Pen
Color Red