Learn More
Corning™ Biocoat™ collagen V, human, 0.25mg
Product Code. 11533560 Shop All Corning Products
Click to view available options
Quantity:
0.25 mg
Unit Size:
250µg
Product Code. 11533560
Brand: Corning™ 354246
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Corning Collagen V, human, is used as a thin coating on tissue-culture surfaces to study collagen V effects on cell behaviour.
  • Sourced from human placenta. NOTE: Source material tested for hepatitis B antigen and HIV-1 antibody
  • Electrophoretically homogeneous (SDS-PAGE)
  • Tested and found negative for bacteria, fungi, and mycoplasma
  • Stable for at least three months at -20°C. Avoid multiple freeze-thaws
  • Recommended concentration - 1-5 μg/cm2 of growth surface depending on cell type
  • Formulation - frozen in 10mM acetic acid
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.25 mg