missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Biocoat™ epidermal growth factor (EGF), mouse natural (receptor grade), 500μg

Product Code. 11553610 Shop All Corning Products
This item is not returnable. View return policy

Product Code. 11553610

Brand: Corning™ 356010

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Corning Epidermal Growth Factor (EGF), mouse natural (receptor grade),500μg, is a low-molecular-weight mitogenic protein that stimulates proliferation of a wide variety of cell types in vitro.
  • Formulation: lyophilised from ammonium acetate, carrier-free; reconstitute in dH2O
  • Stable for at least three months at 2° to 8°C (lyophilised) or one month solubilised at -20°C
  • Do not store in frost-free freezer. Avoid multiple freeze/thaws.
  • Purity greater than 98% by SDS-PAGE and pH 9.9 PAGE
  • Tested for ability to stimulate proliferation of human foreskin fibroblasts in serum-containing medium
  • Filtered (0.2μm membrane) and tested and found negative for bacteria, fungi, and mycoplasma
  • Sourced from male mouse submaxillary glands
  • Molecular Weight 6.1 kD
  • Purified by BioGel P10/DEAE chromatography
  • Recommended concentration: approximately 5ng/mL medium depending on cell type
TRUSTED_SUSTAINABILITY

Specifications

Purity or Quality Grade >98% by SDS-PAGE and pH 9.9 PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.