missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Biocoat™ Fibronectin 175cm2 Rectangular Straight Neck Cell Culture Flask
GREENER_CHOICE

Product Code. 10192761
Change view
Click to view available options
Height (Metric):
175 cm2
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
10192761 175 cm2 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10192761 Supplier Falcon™ Supplier No. 354526

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells

Tested for their ability to promote attachment and spreading of BHK-21 hamster kidney cells.
  • Matrix components are applied as an even, optically clear coating
  • Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
  • All lots tested and found negative for bacteria and fungi
  • Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance All lots tested and found negative for bacteria and fungi
Color Optically clear
Description Straight neck cell culture flask with plug seal cap
For Use With (Application) Promotes cell attachment and spreading and improves the survival of primary cells in culture
Height (Metric) 175 cm2
Neck Style Straight neck
Shape Rectangular
Surface Coating Fibronectin-coated
Temperature Range (Metric) Store between 2°C and 8°C. Do not freeze
Graduated Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.