missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Laminin Rectangular Canted Neck Cell Culture Flask w/Plug Seal Cap

Description
- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
These products are for research use only and are not intended for diagnostic purposes.
For adhesion, growth and differentiation of neuronal and transfected cell lines
Specifications
Specifications
| Capacity (English) | 8.45 oz. |
| Closure Material | Phenolic |
| Capacity (Metric) | 250 mL |
| For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
| Neck Style | Canted |
| No. per Case | 10/Cs. |
| No. per Pack | 5/Pk. |
| Shape | Rectangular |
| Surface Coating | Laminin |
| Sterility | Sterile |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?