missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biocytin Hydrazide

Product Code. 11514167
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11514167 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11514167 Supplier Invitrogen™ Supplier No. B1603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

biocytin hydrazide

Biocytin hydrazide an be used as microinjectable anterograde and transneuronal tracers that is fixable with aldehyde-based fixatives. Additionally, the primary aliphatic amine of this molecule can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Biocytin Hydrazide
Quantity 25 mg
Content And Storage Store at room temperature.
Reagent Type Biocytins
Shipping Condition Room Temperature
Label Type Biotin & Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.