missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ Biohazard Bags, Irradiated
GREENER_CHOICE

Product Code. 18886070 Shop All Micronova Products
Change view
Click to view available options
Color:
Red
Yellow
Capacity (Metric):
120 L
200 L
Unit Size:
Each
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (Metric) Unit Size
18886070 Red 200 L Each
17843818 Yellow 120 L Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18886070 Supplier Micronova™ Supplier No. BL44060REIRBB
  • ONLINE EXCLUSIVE PRICE
    534.00 €
    500.03 € / Pack of 1
    Save 33.97 €
    6% Off
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made from FDA-compliant LDPE, each Biohazard Bag displays a large biohazard symbol and warning message. Supplied irradiated [minimum dose: 25 kGy (SAL: 10-6)] they are ideal for aseptic suites in cell therapy and related disciplines. Available flat packed in groups of 5 or on a plastic core roll.

  • Product Type: Biohazard Bag
  • Material: LDPE
  • Thickness: 4 mm
  • Sterile: Yes
  • Sterilization Method: Irradiation [Minimum Dose: 25 kGy (SAL: 10-6)]

Specifications

Capacity (Metric) 200 L
Certifications/Compliance FDA: Regulation 21 CFR, 177.1520, 3.1; DOT: ASTM D1709 and ASTM D1922; Manufacturing: ISO 9001; Controlled Environments: ISO 5 - 9 Cleanrooms
Closure Type None
Design Biohazard Symbol and Warning Printed on Each Bag
Capacity (English) 55 gal.
Color Red
Dimensions (L x W) 40 x 60 in.
Disposable Single-use
Quantity 50/Cs.
Shape No Pleats at Bottom
Sterility Sterile
Autoclavable Not Autoclavable
For Use With (Application) Biohazardous Waste Disposal
Thickness 4 mm
Packaging 5 Liners/Double Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.