missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Biohazard Disposal Bags
GREENER_CHOICE

Product Code. 16330676 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Product Type:
Bag Holder
Benchtop Biohazard Disposal Bag
Capacity (Metric):
2 L
9 L
19 L
40 L
Unit Size:
Each
Pack of 200
Pack of 500
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Capacity (Metric) Unit Size
16330676 Benchtop Biohazard Disposal Bag 40 L Pack of 200
15519083 Benchtop Biohazard Disposal Bag 2 L Pack of 500
16320676 Benchtop Biohazard Disposal Bag 9 L Pack of 200
15529083 Benchtop Biohazard Disposal Bag 19 L Pack of 200
16300676 Bag Holder 2 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16330676 Supplier Heathrow Scientific™ Supplier No. HS10323
  • ONLINE EXCLUSIVE PRICE
    349.00 €
    313.20 € / Pack of 200
    Save 35.80 €
    10% Off
View all promotions
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag

Specifications

Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Application) Biohazard Disposal
For Use With (Equipment) Autoclave
Material Polypropylene
Sterility Non-sterile
No. per Case 500 Pk.
Outer Packaging Box
Thickness (English) 63 mil
Dimensions (L x W) 25 x 33 in.
Dimensions (L x W x H) 25 x 33 x 0.06 in.
Length (English) 25 in.
Width (English) 33 in.
Height (English) 0.06 in.
Height (Metric) 0.15 cm
Weight (English) 0.08 lb.
Weight (Metric) 0.04 kg
Product Type Benchtop Biohazard Disposal Bag
Capacity (English) 10.6 gal.
Capacity (Metric) 40 L
Length (Metric) 63.5 cm
Width (Metric) 83.8 cm
Thickness (Metric) 1.6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.