missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Disposal Bags with or without Warning Label

Product Code. 11392144
Change view
Click to view available options
Dimensions (L x W):
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Color:
Clear
Translucent
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Color unitSize
11392144 24 x 12 in. (61 x 30 cm) Translucent Pack of 100
11352154 24 x 12 in. (61 x 30 cm) Clear Pack of 100
11302154 30 x 24 in. (76 x 61 cm) Translucent Pack of 100
11362154 30 x 24 in. (76 x 61 cm) Clear Pack of 100
11312154 36 x 24 in. (91 x 61 cm) Translucent Pack of 100
11372154 36 x 24 in. (91 x 61 cm) Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11392144 Supplier Bel-Art™ Supplier No. F131600005
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes

  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners

Specifications

Capacity (Metric) 15 L
Certifications/Compliance ASTM D1709-98
Closure Type Zip Tie
Design Non-printed without Sterilization Patch
Capacity (English) 3 gal.
Color Translucent
Dimensions (L x W) 24 x 12 in. (61 x 30 cm)
Disposable Single-use
Length (Metric) 61 cm
Sterility Non-sterile
Width (Metric) 30 cm
Autoclavable Autoclavable
For Use With (Application) Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Width (English) 12 in.
Length (English) 24 in.
Thickness 1.5 mil (0.03 mm)
Material Polypropylene
Product Type Biohazard Bag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.