missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ X6 Biohazard Keeper, Floor Model

Product Code. 30804793 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Dimensions (L x W x H):
30.5 x 30.5 x 68.6 cm (12 x 12 x 27 in.)
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
30804793 30.5 x 30.5 x 68.6 cm (12 x 12 x 27 in.) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30804793 Supplier Heathrow Scientific™ Supplier No. BH2009
  • ONLINE EXCLUSIVE PRICE
    97.50 €
    91.85 € / Pack of 6
    Save 5.65 €
    5% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Efficient biohazard waste disposal

  • The Biohazard Keeper Floor Model offers a sturdy and economical solution for collecting non-sharp biohazardous lab waste. Designed for easy use and odor control
  • Wide-mouth Reclose-A/Remove-A-Lid for easy disposal and odor reduction
  • Durable corrugated construction with a 2 mil red plastic liner for leak resistance
  • Ideal for petri dishes, tubes, plates, pipets, and specimen cups
  • Clearly marked with orange biohazard symbols and bilingual safety warnings

Specifications

Capacity (Metric) 63 L
Capacity (English) 16.64 gal.
Color White, Blue
Dimensions (L x W x H) 30.5 x 30.5 x 68.6 cm (12 x 12 x 27 in.)
Disposable Single-use
Model Floor Model
Quantity 6/Cs.
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Corrugated Cardboard
Product Type Biohazard Waste Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.