missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heathrow Scientific X50 Biohazard Pipet Keeper Container
Shop All Heathrow Scientific Products

Click to view available options
Dimensions (L x W x H):
7.62 x 7.62 x 39.37 cm (3 x 3 x 15.5 in.)
Unit Size:
Pack of 50
Description
- Pipet keeper provides a reliable solution for collecting lab contaminants while keeping workstations tidy and compliant
- Holds biohazardous waste up to 15 in. length
- Sturdy paperboard construction with visible biohazard symbols
- Self-locking top and re-closable flaps for secure containment
- Easily disposable after autoclaving per regulations
- Cost-effective and user-friendly design
Specifications
Specifications
| Capacity (English) | 2300 cc |
| Color | White, Orange, Blue |
| Dimensions (L x W x H) | 7.62 x 7.62 x 39.37 cm (3 x 3 x 15.5 in.) |
| Quantity | 50/Cs. |
| Autoclavable | Autoclavable |
| Material | Plastic-lined Paperboard |
| Product Type | Pipet Keeper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction