missing translation for 'onlineSavingsMsg'
Learn More

Vilber Louvert Biolink™ BLX UV Crosslinker

Product Code. 11840520
Change view
Click to view available options
Wavelength:
254 nm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength Unit Size
11840520 254 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11840520 Supplier Vilber Louvert Supplier No. 611110811

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rich robust UV crosslinker ideal for the removal of PCR contaminants.

  • Bio-Link Irradiator 254nm
  • Fixation of nucleic acids on membranes
  • DNA photolysis in agarose gel
  • Selection of the RecA mutation
  • Partial digestion by formation of thimine dimers
  • Removal of PCR contaminants
  • UV Sterilization
  • 2 programming modes: energy or time
  • For each mode, 9 programs in memory and one free programming input
  • Storage of the last irradiation setpoint
  • 14mm digital LED display
  • UV-treated control window
  • Stopping the irradiation when the door is opened
  • Control UV illumination is continuously monitored by a radiometer
  • Irradiation stops as soon as the amount of programmed energy has been received by the membrane, thus ensuring better reproducibility

Specifications

Description Vilbert Lourmat Biolink BLX UV Crosslinker
Wavelength 254 nm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.