Learn More
Sperian™ Honeywell™ Bionic Faceshield Visors
Product Code. 12635083
Change view
Click to view available options
Description:
Complete faceshield unit
Visor only
Lens Material:
Acetate
Polycarbonate
Lens Coating:
Anti-fog
Uncoated
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Lens Material Lens Coating Unit Size
12635083 Complete faceshield unit Acetate Uncoated Each
12645053 Visor only Acetate Uncoated Each
12655053 Visor only Polycarbonate Anti-fog Each
11826950 Complete faceshield unit Polycarbonate Uncoated Each
12635053 Visor only Polycarbonate - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12635083 Supplier Sperian™ Supplier No. 1011933
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Offering maximum protection in an ergonomic design. Sperian™ Honeywell™ Bionic Faceshield Visors are incredibly adjustable, with 2784 possible wearer positions.
  • Extended top of head and chin protection
  • Large superior-quality visors for maximum visibility
  • Available with either polycarbonate visor for impact resistance or acetate visor for chemical protection
  • Comfortable fit with goggles or respirators
  • Locking mechanism for more complete safety
  • Dielectric design
  • Fully adjustable ratchet headgear and headband
  • Multiple tilt visor positions
  • Extra-soft cushioning on all parts of the shield in contact with the head
  • Breathable extra-soft sweatband is removable for easy washing and replacement
  • Excellent for the agriculture, chemical, forestry, steels and metals and wind energy industries
  • Protects against abrasion, airborne particles, biohazards, chemicals, impact, splashes and more
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 89/686/EEC, EN169 / EN170 / EN166, ISO 9001 / 2000
Lens Coating Uncoated
Description Complete faceshield unit
Lens Tint Clear
Product Type Faceshield
Frame Color Black
Lens Material Acetate