missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bioreactor Probe Assembly for use with Single-Use Bioreactors

Product Code. 30040280 Shop All Thermo Scientific Products
Change view
Click to view available options
Includes:
Aseptiquick G Connector
Kleenpak Connector
Unit Size:
Pack of 10
Product Code. Includes unitSize
30040280 Kleenpak Connector Pack of 10
30040274 Aseptiquick G Connector Pack of 10
2 options
This item is not returnable. View return policy

Product Code. 30040280

Brand: Thermo Scientific™ SH30720.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bioreactor Probe Assembly for HyPerforma™ and DynaDrive™ Single-use Bioreactors.

  • The Thermo Scientific™ Bioreactor Probe Assembly (nonsterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ Single-Use Bioreactors.

Specifications

Autoclavable Autoclavable
Material Silicone
For Use With HyPerforma™ Single-use Bioreactors, DynaDrive™ Single-use Bioreactors
Includes Kleenpak Connector
Color Clear
Product Type Probe Assembly
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.