Learn More
Agilent BioTek™ BioSpa 8 Left Pan Accessory
GREENER_CHOICE
Product Code. 18440800 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
BioSpa 8 Automated Incubator
Unit Size:
1 unit
Product Code. 18440800
Brand: Agilent BioTek™ 1410019
For use with BioSpa 8 Automated Incubator. Requires Cytation C10 Anti-vibration Base and BioSpa 8 Left Pan Accessory, both purchased separately.
  • For use with BioSpa 8 Automated Incubator.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Liquid Protection Tray
For Use With (Equipment) BioSpa 8 Automated Incubator
Sterility Non-sterile