Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
Shop All Agilent BioTek Products
Click to view available options
Volume (Metric):
10 to 3000 μL
5 to 2500 μL
500 nL to 50 μL
8 to 30,000 μL
Increments:
1 μL
10 μL
5 μL
Not Applicable
Tip Type:
Plastic
Plastic, with large bore
Steel
Unit Size:
Each
Description
For use with MultiFlo FX Microplate Dispenser RAD peri cassette
Specifications
Specifications
| Volume (Metric) | 5 to 2500 μL |
| For Use With (Equipment) | MultiFlo FX Microplate Dispenser |
| Increments | 5 μL |
| Tip Type | Steel |