Learn More
Agilent BioTek Accessories For MultiFlo FX Microplate Dispenser RAD peri cassette
GREENER_CHOICE
Product Code. 16592092 Shop All Agilent BioTek Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
10 to 3000 μL
5 to 2500 μL
500 nL to 50 μL
8 to 30,000 μL
Increments:
1 μL
10 μL
5 μL
Not Applicable
Tip Type:
Plastic
Plastic, with large bore
Steel
missing translation for 'unitSize'
Each
Product Code. 16592092
missing translation for 'mfr': Agilent BioTek 1260017
Only null left
Add to basket
Add to basket
Accurate and precise peristaltic pump dispensing.
For use with MultiFlo FX Microplate Dispenser RAD peri cassette
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 500 nL to 50 μL
For Use With (Equipment) MultiFlo FX Microplate Dispenser
Increments 1 μL
Tip Type Plastic