missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Biotin Alkyne (PEG4 carboxamide-Propargyl Biotin)

Product Code. 10186943
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10186943

Brand: Molecular Probes™ B10185

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

biotin alkyne (PEG4 carboxamide-propargyl biotin)

The hapten, biotin azide is reactive with terminal via a copper-catalyzed click reaction. Biotin can be subsequently detected with streptavidin, avidin or NeutrAvidin™ biotin-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 1 mg
Product Type Biotin Alkyne
Molecular Weight (g/mol) 528.66 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C and keep desiccated.
Chemical Reactivity Azide
Reactive Group Alkyne
Shipping Condition Room Temperature
Reactive Moiety Alkyne
Label or Dye Biotin
Label Type Biotin and Other Haptens
Color None
Product Line Molecular Probes™
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.