missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Biotin-PEG3-Azide

Product Code. 18756335
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18756335 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18756335 Supplier R&D Systems™ Supplier No. 7524/50
  • ONLINE EXCLUSIVE PRICE
    237.00 €
    223.65 € / 50mg
    Save 13.35 €
    5% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Biotin with PEG linker and reactive click handle

Biotin-PEG3-Azide is a biotinylation reagent for labeling alkyne-containing biomolecules either via Cu(I)-catalyzed Azide-Alkyne Click Chemistry reaction (CuAAC) or via Cu(I)-free Strain-Promoted Alkyne-Azide Click Chemistry (SPAAC) reaction with cyclooctyne derivatives.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 AS,4 S,6 aR)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1 H-thieno[3,4-d]imidazole-4-pentanamide
CAS 875770-34-6
Quantity 50 mg
Product Line Tocris
Target General Laboratory Research Reagent
Molecular Formula C18H32N6O5S
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.