missing translation for 'onlineSavingsMsg'
Learn More

Biotin-TEG Azide, TRC

Product Code. 30134545
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134545 25 mg 25mg
30134992 100 mg 100mg
30008724 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134545 Supplier Toronto Research Chemicals Supplier No. TRCB39160025MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875770-34-6
Molecular Formula C18H32N6O5S
Molecular Weight (g/mol) 444.55
Synonym Azide-PEG3-biotin, N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, (3aS,4S,6aR)-N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-[2-[2-(2-azidoethoxy)ethoxy]ethoxy]ethyl]pentanamide
SMILES [N-]=[N+]=NCCOCCOCCOCCNC(CCCC[C@H]1[C@@H]2[C@@H](NC(N2)=O)CS1)=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H32N6O5S/c19-24-21-6-8-28-10-12-29-11-9-27-7-5-20-16(25)4-2-1-3-15-17-14(13-30-15)22-18(26)23-17/h14-15,17H,1-13H2,(H,20,25)(H2,22,23,26)/t14-,15-,17-/m0/s1
Formula Weight 444.2155
Chemical Name or Material Biotin-TEG Azide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.