missing translation for 'onlineSavingsMsg'
Learn More

Biotin, TRC

Product Code. 17947564
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17947564 500 mg 500mg
17979854 1 g 1g
17908604 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17947564 Supplier Toronto Research Chemicals Supplier No. TRCB389040500MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10 H16 N2 O3 S
Molecular Weight (g/mol) 244.31
Synonym 1H-Thieno[3,4-d]imidazole-4-pentanoic acid, hexahydro-2-oxo-, [3aS-(3aα,4β,6aα)]-, Biotin (8CI), (3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanoic acid, (+)-Biotin, (+)-cis-Hexahydro-2-oxo-1H-thieno[3,4]imidazole-4-valeric acid, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Biodermatin, Bioepiderm, Bios II, Coenzyme R, D(+)-Biotin, D-Biotin, Factor S, Factor S (vitamin), Lutavit H2, MD 1003, Medebiotin, Meribin, NSC 63865, Rovimix H 2, Vitamin B7, Vitamin H, Vitamin I, cis-(+)-Tetrahydro-2-oxothieno[3,4]imidazoline-4-valeric acid, d-Biotin
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoic acid
SMILES OC(=O)CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1
Formula Weight 244.0882
Percent Purity >95%
Chemical Name or Material Biotin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.