missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-Ubiquitin

Product Code. 11555016
Change view
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11555016 100 μg 100µg
11545016 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11555016 Supplier Invitrogen™ Supplier No. PV4380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination.

Biotin-labeled ubiquitin for use as a general research tool, or in high throughput assays to monitor protein ubiquitination. Due to the method in labeling the ubiquitin protein, all of the lysines with the protein are available for further conjugation to form poly-ubiquitin chains.

For use in combination with other LanthaScreen™ products including LanthaScreen™ Tb-Ubiquitin (Cat. No. PV4375), Fluorescein-Ubiquitin (PV4377), or a LanthaScreen™ antibody such as Tb-anti-biotin (PV4749).

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1.19mg/mL
For Use With (Application) Proteasome Assay, Drug Discovery and Development, Epigenetics Assay, Pharma and Biopharma, Proteasome Biology, Target and Lead Identification and Validation
Molecular Weight (g/mol) 9.2kDa
Name Biotin-Ubiquitin
Quantity 100 μg
Storage Requirements –20°C
Regulatory Status For research use only. Not for use in diagnostic procedures
Packaging 1 Each
Common Name Biotin-Ubiquitin
Includes Biotin labeled ubiquitin in HEPES buffered saline
Product Type Biotin-Ubiquitin
Conjugate Biotin
Content And Storage Biotin labeled ubiquitin in HEPES buffered saline. Store at -20°C.
Detection Method Fluorescence
Excitation/Emission 340/490, 520 nm
For Use With (Equipment) Microplate Reader
Label or Dye Biotin
Label Type Biotin & Other Haptens
Readout End Point
Shipping Condition Dry Ice
Substrate Properties Protein-Based Substrate
Technique TR-FRET
Form Liquid
Purity or Quality Grade ≥90% by SDS-PAGE
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.